Skip to Content
Merck

Skip To

AB2043

Anti-Elastin Antibody

Chemicon®, from rabbit

Synonym(s):

Anti-Elastin, Anti-Elastin Antibody, Elastin Detection Antibody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 12,100.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA, IHC, RIA
Citations:
1

CZK 12,100.00


Available to ship TODAYDetails


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

should not react with

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable, immunohistochemistry: suitable, radioimmunoassay: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ELN(2006)

Immunogen

Elastin extracted and purified from human skin.

Application

Detect Elastin using this Anti-Elastin Antibody validated for use in ELISA, RIA, IH.
Immunohistochemistry: 1:80 dilution for immunofluorescent staining of frozen human skin and liver tissues.

Radioimmunoassay

ELISA

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Biochem/physiol Actions

Antibody shows less than 0.1% reactivity with human Collagen I, II, III, IV, V and human Fibronectin as well as Rat Elastin by radioimmunoassay.

Physical form

Format: Purified
The purified antibody is supplied in a buffer containing a mixture of 0.1M Citrate, 0.1M potassium phosphate, at a pH of 7.2-7.4 and 10 ul/ml of antibiotics and antimycotics

Preparation Note

Maintain frozen at -20°C for up to 12 months from date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Control tissue: human skin or liver

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MAB2503HPA056941E4013
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

mouse

biological source

rabbit

biological source

mouse

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

200

antibody form

purified antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

ascites fluid

technique(s)

ELISA: suitable, radioimmunoassay: suitable, immunohistochemistry: suitable

technique(s)

ELISA: suitable, immunofluorescence: suitable, immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:50-1:200

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:5,000 using sheep arterial tissue, microarray: suitable

clone

polyclonal

clone

10B8, monoclonal

clone

polyclonal

clone

BA-4, monoclonal


Still not finding the right product?

Explore all of our products under Anti-Elastin Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters