Skip to Content
Merck
All Photos(1)

Documents

AB1559P

Sigma-Aldrich

Anti-Adenosine A2a Receptor Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

pig, bovine, canine, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ADORA2A(135)

Specificity

Adenosine A2a receptor. Recognizes a band of approximately 45 kDa in human (coronary artery), and rat, porcine and bovine striatal membranes.

Immunogen

Amino acid sequence near the 4th C-terminal intracellular domain (30 amino acids) of the cloned canine A2a receptor. The A2a sequence selected is not present in A1, A2b and A3 receptors. Control peptide is available for this product (Catalog Number AG285).

Application

Anti-Adenosine A2a Receptor Antibody is an antibody against Adenosine A2a Receptor for use in ELISA, IC, IH & WB.
Immunohistochemistry: 10-50 μg/mL on frozen tissue sections

Western blot: 1-10 μg/mL (Chemiluminescence technique), 40-100 μg membrane proteins/lane of striatal membranes. Higher concentrations of the antibody may be necessary if colorimetric detection or milk-based antibody diluent are used.

Immunocytochemistry: Caufield, 2003. Light 2% PFA fixation 5-10 minutes --long fixations lower staining intensity.

ELISA: 1:10,000-1:50,000 (50-100 ng A2aR peptide (Cat. Number AG285)/well)

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Circadian Rhythm & Sleep

Physical form

Affinity purified immunoglobulin. Liquid in PBS with 0.1% BSA and 0.05% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gemma Navarro et al.
Nature communications, 9(1), 1242-1242 (2018-03-30)
G protein-coupled receptors (GPCRs), G proteins and adenylyl cyclase (AC) comprise one of the most studied transmembrane cell signaling pathways. However, it is unknown whether the ligand-dependent interactions between these signaling molecules are based on random collisions or the rearrangement

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service