Skip to Content
Merck
All Photos(1)

Documents

AB1532SP

Sigma-Aldrich

Anti-Neurotrophin 3 Antibody

Chemicon®, from rabbit

Synonym(s):

NT3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, chicken, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NTF3(4908)

Specificity

Neurotrophin 3 (NT3). By dot blot, less than 0.1% cross-reactivity against mouse NGF, recombinant human BDNF or neurotrophin 4.

Immunogen

An 11 amino acid peptide corresponding to the amino-terminal of mouse Neurotrophin 3 (NT3) coupled to BSA. The sequence is H-YAEHKSHRGEY-NH2.

Application

Immunohistochemistry: 1-10 μg/mL (see suggested protocol).

Immunoblotting: 1-10 μg/mL

ELISA: 1-10 μg/mL

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins
This Anti-Neurotrophin 3 Antibody is validated for use in ELISA, WB, IH for the detection of Neurotrophin 3.

Physical form

Format: Purified
Protein A purified immunoglobulin. Lyophilized from PBS, no preservatives. Reconstitute with 500 μL of sterile distilled water (1 mg/mL).

Storage and Stability

Maintain at -20 to -70°C in undiluted aliquots for up to one year. Avoid repeated freeze/thaw cycles. Glycerol (1:1) can be added for additional stability.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leandro Viçosa Bonetti et al.
Muscle & nerve, 53(4), 617-625 (2015-09-01)
Numerous rehabilitation treatments have been shown to be useful for peripheral and central restoration after (PNI). After sciatic nerve crush, we investigated 4 weeks of endurance training (ET) and balance and coordination training (BCT) with sciatic function index, hind-paw stride
Dong Hoon Hwang et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(38), 12788-12800 (2014-09-19)
Combining cell transplantation with activity-based rehabilitation is a promising therapeutic approach for spinal cord repair. The present study was designed to investigate potential interactions between the transplantation (TP) of neural stem cells (NSCs) obtained at embryonic day 14 and treadmill

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service