Skip to Content
Merck

Skip To

8.56116

Fmoc-Ser(tBu)-Wang resin LL (100-200 mesh)

Novabiochem®

Synonym(s):

Fmoc-Serine Wang Resin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 g
Please contact Customer Service for Availability
CZK 2,050.00
5 g
Please contact Customer Service for Availability
CZK 6,240.00

About This Item

NACRES:
NA.22
UNSPSC Code:
12352209

CZK 2,050.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-30°C

General description

Pre-loaded resin for synthesis of peptide acids containing a C-terminal serine amino-acid residue by Fmoc SPPS. Prepared from a low load Wang resin, to help facilitate the synthesis of longer peptides.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Application

Applications of Fmoc-Ser(tBu)-Wang resin LL (100-200 mesh) include:
  • In the preparation of a peptide thioester intermediate for use in the solid-phase synthesis of peptide and glycopeptide thioesters using a side-chain-anchoring strategy.
  • As a solid support in the synthesis of a peptide substrate for use in a reaction to prepare disubstituted pyrazino-oxazine derivatives.
  • In the synthesis of a peptide intermediate for use in the solid-phase synthesis of tetramic acid derivatives, which have potential applications as pharmaceuticals.

Other Notes

Replaces: 04-12-2086

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.561188.561178.56114
reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

functional group

Fmoc

functional group

Boc, Fmoc

functional group

Fmoc

functional group

Fmoc

Quality Level

400

Quality Level

400

Quality Level

400

Quality Level

400

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
856116000104027269119898
856116000504027269119904

Questions

Reviews

No rating value

Active Filters