Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 5 g | Please contact Customer Service for Availability | CZK 7,870.00 | |
| 25 g | Please contact Customer Service for Availability | CZK 31,400.00 |
About This Item
Quality Segment
product line
Novabiochem®
form
beads
reaction suitability
reaction type: solution phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
storage temp.
2-8°C
General description
Literature references
[1] E. Valeur, et al. (2005) Chem. Commun., 1164.
[2] Y. Kiso, et al. (1973) Chem. Pharm. Bull., 21, 2507.
[3] J. R. Vaughan (1951) J. Am. Chem. Soc., 73, 3547.
[4] E. Valeur & M. Bradley, unpublished results.
Analysis Note
Appearance of substance (visual): beads
Loading (determined by HPLC after reaction of 3-phenylpropionic acid with cyclohexlamine): 1.3 - 1.9 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly (styrene-1 % DVB) 200 - 400 mesh.
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| reaction suitability reaction type: solution phase peptide synthesis | reaction suitability reaction type: Coupling Reactions | reaction suitability reaction type: solution phase peptide synthesis, reactivity: alcohol reactive | reaction suitability reaction type: solution phase peptide synthesis, reactivity: carboxyl reactive |
| product line Novabiochem® | product line Novabiochem® | product line Novabiochem® | product line Novabiochem® |
| Quality Level 200 | Quality Level 300 | Quality Level 200 | Quality Level 200 |
| application(s) peptide synthesis | application(s) peptide synthesis | application(s) peptide synthesis | application(s) peptide synthesis |
| storage temp. 2-8°C | storage temp. -10 to -25°C | storage temp. 2-8°C | storage temp. 2-8°C |
| form beads | form powder | form beads | form beads |
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 8550460025 | 04027269111298 |
| 8550460005 | 04027269111281 |



