Skip to Content
Merck

Skip To

8.53062

Boc-Sar-OH

Novabiochem®

Synonym(s):

Boc-Sar-OH, N-α-t.-Boc-sarcosine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
5 g
Please contact Customer Service for Availability
CZK 565.00
100 g
Please contact Customer Service for Availability
CZK 3,570.00

About This Item

Empirical Formula (Hill Notation):
C8H15NO4
CAS Number:
Molecular Weight:
189.21
UNSPSC Code:
12352209
EC Index Number:
237-306-0
NACRES:
NA.22
MDL number:
eCl@ss:
32160406

CZK 565.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥98% (TLC)

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-30°C

SMILES string

N(CC(=O)[O-])(C)C(=O)OC(C)(C)C

InChI

1S/C8H15NO4/c1-8(2,3)13-7(12)9(4)5-6(10)11/h5H2,1-4H3,(H,10,11)/p-1

InChI key

YRXIMPFOTQVOHG-UHFFFAOYSA-M

General description

Standard building block for introduction of sarcosine amino-acid residues by Boc SPPS.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(157A)): ≥ 98 %
Purity (TLC(0110)): ≥ 98 %
Solubility (1 mmole in 2 ml DMF): clearly soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-0109

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.530038.530278.53070
form

powder

form

powder

form

powder

form

powder

assay

≥98% (TLC)

assay

≥98% (TLC)

assay

≥98% (TLC)

assay

≥98% (TLC)

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

2-30°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

reaction suitability

reaction type: Boc solid-phase peptide synthesis

reaction suitability

reaction type: Boc solid-phase peptide synthesis

reaction suitability

reaction type: Boc solid-phase peptide synthesis

reaction suitability

reaction type: Boc solid-phase peptide synthesis


Still not finding the right product?

Explore all of our products under Boc-Sar-OH


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Unnatural amino acids, the non-proteinogenic amino acids that either occur naturally or are chemically synthesized, are becoming more and more important as tools for modern drug discovery research.






Global Trade Item Number

SKUGTIN
853062000504027269098476
853062002504027269098483
853062010004027269098490

Questions

Reviews

No rating value

Active Filters