Skip to Content
Merck

Skip To

8.51071

Mca-OH

Novabiochem®

Synonym(s):

Mca-OH, 7-Methoxycoumarin-4-acetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 g
Please contact Customer Service for Availability
CZK 3,220.00
5 g
Please contact Customer Service for Availability
CZK 12,800.00

About This Item

Empirical Formula (Hill Notation):
C12H10O5
CAS Number:
Molecular Weight:
234.20
UNSPSC Code:
12352005
NACRES:
NA.22
MDL number:
Assay:
≥95% (TLC), ≥95.0% (HPLC)
Form:
powder

CZK 3,220.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

product line

Novabiochem®

assay

≥95% (TLC), ≥95.0% (HPLC)

form

powder

reaction suitability

reagent type: fluorescent-labelling reagent

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

[o]1c2c(c(c[c]1=O)CC(=O)O)ccc(c2)OC

InChI

1S/C12H10O5/c1-16-8-2-3-9-7(4-11(13)14)5-12(15)17-10(9)6-8/h2-3,5-6H,4H2,1H3,(H,13,14)

InChI key

ZEKAXIFHLIITGV-UHFFFAOYSA-N

General description

7-Methoxycoumarin (Mca) fluoresces at 405 nm when stimulated at 340 nm, and is most commonly used in conjunction with 2,4-dinitrophenyl quenching group.

Associated Protocols and Technical Articles
Chromogenic Labeling of Synthetic Peptides

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(CMA1)): ≥ 95 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 01-63-0111

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.520958.52309SML2944
assay

≥95% (TLC), ≥95.0% (HPLC)

assay

≥97% (TLC), ≥97.0% (HPLC)

assay

≥95.0% (HPLC), ≥98% (TLC)

assay

≥97% (HPLC)

reaction suitability

reagent type: fluorescent-labelling reagent

reaction suitability

-

reaction suitability

-

reaction suitability

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

form

powder

form

powder

form

powder

form

film or powder

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

-


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Chromogenic and fluorogenic derivatives are invaluable tools for biochemistry, having numerous applications in enzymology, protein chemistry, immunology and histochemistry.





Global Trade Item Number

SKUGTIN
851071000504027269083618
851071000104027269083601

Questions

Reviews

No rating value

Active Filters