Skip to Content
Merck

Skip To

8.44009

Octamethyltrisiloxane

for synthesis

Synonym(s):

Octamethyltrisiloxane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 L
Please contact Customer Service for Availability
CZK 6,320.00
CZK 4,740.00

About This Item

Linear Formula:
[(CH3)3SiO]2Si(CH3)2
CAS Number:
Molecular Weight:
236.53
UNSPSC Code:
12352103
NACRES:
NA.22
EC Index Number:
203-497-4
MDL number:
Beilstein/REAXYS Number:
1753063

CZK 4,740.00

List PriceCZK 6,320.00Save 25%

Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

>1 (vs air)

Quality Level

form

liquid

refractive index

n20/D 1.384 (lit.)

bp

153 °C (lit.)

mp

−82 °C (lit.)

transition temp

flash point 29 °C

density

0.82 g/cm3 at 25 °C, 0.82 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

C[Si](C)(C)O[Si](C)(C)O[Si](C)(C)C

InChI

1S/C8H24O2Si3/c1-11(2,3)9-13(7,8)10-12(4,5)6/h1-8H3

InChI key

CXQXSVUQTKDNFP-UHFFFAOYSA-N

General description

for synthesis

Application

Octamethyltrisiloxane for synthesis. CAS 107-51-7, molar mass 236.53 g/mol.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
2357094693191.00163
density

0.82 g/cm3 at 25 °C, 0.82 g/mL at 25 °C (lit.)

density

0.82 g/mL at 25 °C (lit.)

density

0.82 g/mL at 25 °C (lit.)

density

2.46 g/cm3 at 20 °C

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

100

mp

−82 °C (lit.)

mp

−82 °C (lit.)

mp

−82 °C (lit.)

mp

450 °C

storage temp.

2-30°C

storage temp.

-

storage temp.

-

storage temp.

2-30°C

refractive index

n20/D 1.384 (lit.)

refractive index

n20/D 1.384 (lit.)

refractive index

n20/D 1.384 (lit.)

refractive index

-

bp

153 °C (lit.)

bp

153 °C (lit.)

bp

153 °C (lit.)

bp

-


Still not finding the right product?

Explore all of our products under Octamethyltrisiloxane


pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

84.2 °F - (External MSDS)

flash_point_c

29 °C - (External MSDS)



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
844009010004027269701765
844009100004027269701772

Questions

Reviews

No rating value

Active Filters