Skip to Content
Merck

Skip To

8.44002

Dimethylsuccinate

for synthesis

Synonym(s):

Dimethylsuccinate, Succinic acid dimethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 L
Please contact Customer Service for Availability
CZK 2,030.00

About This Item

CAS Number:
Molecular Weight:
146.14
UNSPSC Code:
12352108
EC Index Number:
203-419-9
NACRES:
NA.22
MDL number:
Form:
liquid

CZK 2,030.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.2 hPa ( 20 °C)

Quality Segment

form

liquid

autoignition temp.

365 °C

potency

>5000 mg/kg LD50, oral (Rat), >5000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.0-8.5 % (v/v)

bp

196 °C/1013 hPa

mp

18-19 °C

transition temp

flash point 94 °C

solubility

75 g/L

density

1.12 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C)C(=O)CCC(=O)OC

InChI

1S/C6H10O4/c1-9-5(7)3-4-6(8)10-2/h3-4H2,1-2H3

InChI key

MUXOBHXGJLMRAB-UHFFFAOYSA-N

General description

for synthesis

Application

Dimethylsuccinate for synthesis. CAS 106-65-0, molar mass 146.14 g/mol.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.201508.400998.00680
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

solubility

75 g/L

solubility

75 g/L

solubility

-

solubility

-

density

1.12 g/cm3 at 20 °C

density

1.12 g/cm3 at 20 °C

density

0.89 g/cm3 at 20 °C

density

1.04 g/cm3 at 20 °C

form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

storage temp.

2-30°C

mp

18-19 °C

mp

18-19 °C

mp

-75 °C

mp

-20 °C


Still not finding the right product?

Explore all of our products under Dimethylsuccinate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_c

94 °C

flash_point_f

201.2 °F



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters