Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
5 mL | Please contact Customer Service for Availability | CZK 689.00 |
250 mL | Please contact Customer Service for Availability | CZK 830.00 |
1 L | Please contact Customer Service for Availability | CZK 2,610.00 |
About This Item
vapor pressure
1.33 hPa ( 55 °C)
Quality Segment
assay
≥99% (GC)
form
liquid
potency
8530 mg/kg LD50, oral (Rat), >5000 mg/kg LD50, skin (Rat)
bp
215-217 °C/1013 hPa
mp
-20 °C
transition temp
flash point 80 °C
density
1.04 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
O(CC)C(=O)CCC(=O)OCC
InChI
1S/C8H14O4/c1-3-11-7(9)5-6-8(10)12-4-2/h3-6H2,1-2H3
InChI key
DKMROQRQHGEIOW-UHFFFAOYSA-N
Analysis Note
Density (d 20 °C/ 4 °C): 1.039 - 1.041
Identity (IR): passes test
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥99% (GC) | assay 99% | assay ≥99% | assay ≥99% |
| Quality Level 200 | Quality Level 100 | Quality Level 300, 400 | Quality Level 300, 400 |
| density 1.04 g/cm3 at 20 °C | density 1.047 g/mL at 25 °C (lit.) | density 1.047 g/mL at 25 °C (lit.) | density 1.047 g/mL at 25 °C (lit.) |
| storage temp. 2-30°C | storage temp. - | storage temp. - | storage temp. - |
| mp -20 °C | mp −20 °C (lit.) | mp −20 °C (lit.) | mp −20 °C (lit.) |
| form liquid | form liquid | form - | form - |
Still not finding the right product?
Explore all of our products under Diethyl succinate
Storage Class
10 - Combustible liquids
wgk
WGK 2
flash_point_f
208.4 °F - closed cup
flash_point_c
98 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

