Skip to Content
Merck

Skip To

8.43784

Benzyl propionate

for synthesis

Synonym(s):

Benzyl propionate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
UNSPSC Code:
12352108
EC Index Number:
204-559-3
NACRES:
NA.22
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥99.0% (GC)

form

liquid

potency

3300 mg/kg LD50, oral (Rat), >5000 mg/kg LD50, skin (Rabbit)

bp

222 °C/1013 hPa

transition temp

flash point 96 °C

density

1.029-1.033 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C10H12O2/c1-2-10(11)12-8-9-6-4-3-5-7-9/h3-7H,2,8H2,1H3

InChI key

VHOMAPWVLKRQAZ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.029 - 1.033
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under Benzyl propionate


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

204.8 °F - closed cup

flash_point_c

96 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
843784050004027269984113
843784010004027269984106

Questions

Reviews

No rating value

Active Filters