Skip to Content
Merck

Skip To

8.41864

D-(+)-Glucuronolactone

for synthesis

Synonym(s):

D-(+)-Glucuronolactone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g
Please contact Customer Service for Availability
CZK 993.00
100 g
Please contact Customer Service for Availability
CZK 2,610.00

About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
UNSPSC Code:
12352115
EC Index Number:
251-053-3
NACRES:
NA.22
MDL number:

CZK 993.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97.0% (acidimetric)

form

solid

mp

170-175 °C

storage temp.

2-30°C

SMILES string

O1[C@@H]([C@@H]([C@@H](C1=O)O)O)[C@@H](O)C=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h1-5,8-10H/t2-,3+,4-,5+/m0/s1

InChI key

UYUXSRADSPPKRZ-SKNVOMKLSA-N

Analysis Note

Assay (acidimetric): ≥ 97.0 %
Spec. rotation [α²0/D (c=8 in water): +17.0 - +19.0 °
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
055661.077584440-M
assay

≥97.0% (acidimetric)

assay

≥99.0% (sum of anomers, GC)

assay

-

assay

≥98% (GC, HPLC)

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

100

form

solid

form

-

form

solid

form

solid

storage temp.

2-30°C

storage temp.

-

storage temp.

15-25°C

storage temp.

10-30°C

mp

170-175 °C

mp

172-175 °C (lit.)

mp

98-100 °C (lit.)

mp

-


Storage Class

11 - Combustible Solids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters