Skip to Content
Merck

Skip To

4440-M

D-Mannose

Synonym(s):

D-Mannose, D-Mannopyranose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 g

Estimated to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 6,300.00

About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
UNSPSC Code:
12352201
NACRES:
NA.79
MDL number:

CZK 6,300.00


Estimated to ship TODAYDetails


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Merck USA index - 14, 5747

Quality Segment

assay

≥98% (GC, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 2.5 mg/mL

storage temp.

10-30°C

SMILES string

O1C(C(C(C(C1CO)O)O)O)O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2

InChI key

WQZGKKKJIJFFOK-UHFFFAOYSA-N

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M6020M857463579
assay

≥98% (GC, HPLC)

assay

≥99%

assay

≥99% (GC)

assay

≥99.5% (sum of enantiomers, HPLC)

solubility

water: 2.5 mg/mL

solubility

H2O: 50 mg/mL

solubility

water: soluble 10 g/L at 20 °C

solubility

H2O: 1 M, clear, colorless

Quality Level

100

Quality Level

300

Quality Level

200

Quality Level

200

form

solid

form

powder

form

powder

form

solid

storage temp.

10-30°C

storage temp.

-

storage temp.

room temp

storage temp.

room temp

color

white

color

-

color

white to off-white

color

white


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters