Skip to Content
Merck

Skip To

8.22170

Trichlorooctadecylsilane

for synthesis

Synonym(s):

Trichlorooctadecylsilane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mL
Please contact Customer Service for Availability
CZK 3,240.00

About This Item

Empirical Formula (Hill Notation):
C18H37Cl3Si
CAS Number:
Molecular Weight:
387.93
UNSPSC Code:
12352103
EC Index Number:
203-930-7
NACRES:
NA.22
MDL number:

CZK 3,240.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

refractive index

n20/D 1.459

bp

223 °C/13 hPa

transition temp

flash point 89 °C

density

0.98 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Si](Cl)(Cl)(Cl)CCCCCCCCCCCCCCCCCC

InChI

1S/C18H37Cl3Si/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-22(19,20)21/h2-18H2,1H3

InChI key

PYJJCSYBSYXGQQ-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 96.0 %

Refractive Index (20°C): 1.458 - 1.462

Identity (IR): passes test


Still not finding the right product?

Explore all of our products under Trichlorooctadecylsilane


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible, corrosive hazardous materials

wgk

WGK 1

flash_point_f

404.6 °F - closed cup

flash_point_c

207 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library