Skip to Content
Merck

Skip To

8.20975

Phenothiazine

for synthesis

Synonym(s):

Phenothiazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 g
Please contact Customer Service for Availability
CZK 894.00
1 kg
Please contact Customer Service for Availability
CZK 2,650.00
50 kg
Please contact Customer Service for Availability
CZK 50,600.00

About This Item

Empirical Formula (Hill Notation):
C12H9NS
CAS Number:
Molecular Weight:
199.27
UNSPSC Code:
12352005
EC Index Number:
202-196-5
NACRES:
NA.22
MDL number:
Assay:
≥97.0% (HPLC)
Grade:
synthesis grade
Bp:
381 °C/1011 hPa
Vapor pressure:
0.0000647 Pa ( 20 °C)

CZK 894.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Segment

vapor pressure

0.0000647 Pa ( 20 °C)

assay

≥97.0% (HPLC)

form

solid

autoignition temp.

470 °C

potency

>2000 mg/kg LD50, oral (Rat)

dilution

(for synthesis)

pH

6 (20 °C, 10 g/L in H2O, aqueous suspension)

bp

381 °C/1011 hPa

mp

182-185 °C

solubility

0.127 mg/L

density

1.296 g/cm3 at 20 °C

bulk density

650 kg/m3

storage temp.

2-30°C

SMILES string

S1c2c(cccc2)Nc3c1cccc3

InChI

1S/C12H9NS/c1-3-7-11-9(5-1)13-10-6-2-4-8-12(10)14-11/h1-8,13H

InChI key

WJFKNYWRSNBZNX-UHFFFAOYSA-N

Application


  • Structural, electrical, optical, and DFT studies of phenothiazine-based D–ο–A frameworks for dye-sensitized solar cell applications: The study focuses on the utility of phenothiazine in dye-sensitized solar cells emphasizing its electron-donating properties (KS Dhivya, C Senthilkumar, K Karthika, 2024).

Analysis Note

Assay (HPLC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 183 °C
Melting range (upper value): ≤ 187 °C
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
88580916722P14831
Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

grade

synthesis grade

grade

-

grade

-

grade

-

assay

≥97.0% (HPLC)

assay

≥98.0% (GC)

assay

-

assay

≥98%

solubility

0.127 mg/L

solubility

-

solubility

-

solubility

-

form

solid

form

pellets

form

powder

form

-

bp

381 °C/1011 hPa

bp

371 °C (lit.)

bp

-

bp

371 °C (lit.)


Still not finding the right product?

Explore all of our products under Phenothiazine


signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2 Oral

target_organs

Blood

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters