Skip to Content
Merck

Skip To

8.20529

9,10-Diphenylanthracene

for synthesis

Synonym(s):

9,10-Diphenylanthracene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C26H18
CAS Number:
Molecular Weight:
330.42
UNSPSC Code:
12352005
EC Index Number:
216-105-1
NACRES:
NA.22
MDL number:
Assay:
≥98.0% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (HPLC)

form

powder

mp

247-252 °C

storage temp.

2-30°C

SMILES string

c21c(c(c4c(c2c5ccccc5)cccc4)c3ccccc3)cccc1

InChI

1S/C26H18/c1-3-11-19(12-4-1)25-21-15-7-9-17-23(21)26(20-13-5-2-6-14-20)24-18-10-8-16-22(24)25/h1-18H

InChI key

FCNCGHJSNVOIKE-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 247 °C
Melting range (upper value): ≤ 252 °C
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1565078.075648.20970
assay

≥98.0% (HPLC)

assay

≥99%

assay

≥99.0% (HPLC)

assay

≥98.0% (acidimetric)

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

200

storage temp.

2-30°C

storage temp.

-

storage temp.

2-30°C

storage temp.

2-30°C

form

powder

form

-

form

powder

form

powder

mp

247-252 °C

mp

209-212 °C (lit.)

mp

203-206 °C

mp

>350 °C


Still not finding the right product?

Explore all of our products under 9,10-Diphenylanthracene


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters