Skip to Content
Merck
All Photos(1)

Key Documents

8.20395

Sigma-Aldrich

N,N-Diethylaniline

for synthesis

Synonym(s):

N,N-Diethylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N(C2H5)2
CAS Number:
Molecular Weight:
149.23
MDL number:
UNSPSC Code:
12352116
EC Index Number:
202-088-8

vapor pressure

0.2 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

500 °C

potency

610 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rat)

expl. lim.

1.1-5.3 % (v/v)

pH

8 (20 °C, 0.1 g/L in H2O)

bp

217 °C/1013 hPa

mp

-38.8 °C

transition temp

flash point 79 °C

density

0.93 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C10H15N/c1-3-11(4-2)10-8-6-5-7-9-10/h5-9H,3-4H2,1-2H3

InChI key

GGSUCNLOZRCGPQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.933 - 0.934
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Skin Irrit. 2 - STOT RE 2 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service