Skip to Content
Merck

Skip To

8.20255

Carbazole

for synthesis

Synonym(s):

Carbazole, Diphenylenimine, 9-Azafluorene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 g
Please contact Customer Service for Availability
CZK 702.00
250 g
Please contact Customer Service for Availability
CZK 2,060.00

About This Item

Empirical Formula (Hill Notation):
C12H9N
CAS Number:
Molecular Weight:
167.21
UNSPSC Code:
12352005
EC Index Number:
201-696-0
NACRES:
NA.22
MDL number:

CZK 702.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95.0% (GC)

form

powder

potency

>16000 mg/kg LD50, oral (Rat)

bp

355 °C/1013 hPa

mp

244-246 °C

transition temp

flash point 220 °C

bulk density

620 kg/m3

storage temp.

2-30°C

SMILES string

[nH]1c2c(c3c1cccc3)cccc2

InChI

1S/C12H9N/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8,13H

InChI key

UJOBWOGCFQCDNV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Melting range (lower value): ≥ 243 °C
Melting range (upper value): ≤ 246 °C
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
46100C5132PHR1564
form

powder

form

-

form

powder

form

-

assay

≥95.0% (GC)

assay

-

assay

≥95% (GC)

assay

-

storage temp.

2-30°C

storage temp.

-

storage temp.

room temp

storage temp.

2-30°C

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

300

mp

244-246 °C

mp

243-246 °C (lit.)

mp

243-246 °C (lit.)

mp

243-246 °C (lit.)

transition temp

flash point 220 °C

transition temp

-

transition temp

-

transition temp

-


Still not finding the right product?

Explore all of our products under Carbazole


pictograms

Health hazard

signalword

Warning

Hazard Classifications

Aquatic Chronic 4 - Carc. 2 - Muta. 2

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

428.0 °F - closed cup

flash_point_c

220.0 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters