Skip to Content
Merck

Skip To

8.20196

METHYL BROMOACETATE

FOR SYNTHESIS

Synonym(s):

METHYL BROMOACETATE, Bromoacetic acid methyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C3H5BrO2
CAS Number:
Molecular Weight:
152.97
EC Index Number:
202-499-2
UNSPSC Code:
12352108
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

18 hPa ( 50 °C)

Quality Segment

assay

≥97.0% (GC)

bp

144-146 °C/1013 hPa

transition temp

flash point 64 °C

density

1.65 g/cm3 at 20 °C

InChI

1S/C3H5BrO2/c1-6-3(5)2-4/h2H2,1H3

InChI key

YDCHPLOFQATIDS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.653 - 1.658
Identity (IR): passes test


pictograms

CorrosionSkull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1

Storage Class

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

147.2 °F - closed cup

flash_point_c

64 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

96-32-2

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library