Skip to Content
Merck

Skip To

8.18447

2,2′-Methylenebis(4-methyl-6-tert-butylphenol)

for synthesis

Synonym(s):

2,2′-Methylenebis(4-methyl-6-tert-butylphenol), 2,2′-Methylenebis-(6-tert-butyl-4-methyl-phenol), Bis(3-tert-butyl-2-hydroxy-5-methylphenyl)-methan

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(2-(OH)-3-(CH3)3C-5-(CH3)C6H2)2CH2
CAS Number:
Molecular Weight:
340.50
UNSPSC Code:
12352104
EC Index Number:
204-327-1
NACRES:
NA.22
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

<1 hPa

Quality Segment

assay

≥96.0% (GC)

form

crystals

autoignition temp.

350 °C

potency

4880 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rabbit)

pH

4-7 (20 °C in H2O, aqueous suspension)

mp

125-132 °C (Lit.)

transition temp

flash point 185 °C

solubility

practically insoluble 0.007 mg/L

density

1.07 g/cm3 at 20 °C

bulk density

300 kg/m3

storage temp.

2-30°C

SMILES string

Oc1c(cc(cc1Cc2c(c(cc(c2)C)C(C)(C)C)O)C)C(C)(C)C

InChI

1S/C23H32O2/c1-14-9-16(20(24)18(11-14)22(3,4)5)13-17-10-15(2)12-19(21(17)25)23(6,7)8/h9-12,24-25H,13H2,1-8H3

InChI key

KGRVJHAUYBGFFP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Melting range (lower value): ≥ 125 °C
Melting range (upper value): ≤ 132 °C
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
4131438.428578.18449
assay

≥96.0% (GC)

assay

-

assay

≥98.0% (GC)

assay

≥99.0% (HPLC)

storage temp.

2-30°C

storage temp.

-

storage temp.

2-30°C

storage temp.

2-30°C

solubility

practically insoluble 0.007 mg/L

solubility

-

solubility

<0.0001 g/L

solubility

-

density

1.07 g/cm3 at 20 °C

density

-

density

0.99 g/cm3 at 20 °C

density

-

form

crystals

form

powder

form

crystals

form

powder

mp

125-132 °C (Lit.)

mp

119-122 °C (lit.)

mp

153-156 °C

mp

158-162 °C


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

365.0 °F - Pensky-Martens closed cup

flash_point_c

185 °C - Pensky-Martens closed cup

wgk

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

119-47-1

CAS No.

119-47-1

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
818447904504027269032906
818447050004022536436272
818447010004022536436265

Questions

Reviews

No rating value

Active Filters