Skip to Content
Merck

Skip To

8.18199

Sodium ethoxide

(20% solution in ethanol) for synthesis

Synonym(s):

Sodium ethylate, Sodium ethanolate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mL
Please contact Customer Service for Availability
CZK 805.00
CZK 603.75
1 L
Please contact Customer Service for Availability
CZK 2,420.00
50 kg
Please contact Customer Service for Availability
CZK 40,000.00

About This Item

NACRES:
NA.22
UNSPSC Code:
12352107
Form:
liquid
Grade:
synthesis grade

CZK 603.75

List PriceCZK 805.00Save 25%

Please contact Customer Service for Availability
This item is subject to Excise Duty. Please check the cart for more details.

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Segment

vapor pressure

31 hPa ( 20 °C)

form

liquid

autoignition temp.

425 °C

expl. lim.

2.6-19 % (v/v)

pH

11 ( in H2O)

bp

91 °C/1013 hPa

transition temp

flash point 23 °C

density

0.88 g/cm3 at 20 °C

storage temp.

15-25°C

Application

  • Facile synthesis and pesticidal activity of substituted heterocyclic pyridine compounds: Discusses the use of sodium ethoxide in the synthesis of pyridine compounds, demonstrating its utility in the development of new pesticidal agents (Abdel-Raheem et al., 2022).
  • Synthesis and biological activities of some fused pyran derivatives: Sodium ethoxide is employed in the synthesis of biologically active pyran derivatives, illustrating its role in enhancing synthetic efficiency (Shehab and Ghoneim, 2016).
  • Environmentally benign catalyst for chemical depolymerization: Highlights the use of sodium ethoxide as a catalyst in the depolymerization of post-consumer PET waste, presenting an environmentally friendly approach to recycling (Javed et al., 2023).

Analysis Note

Assay (acidimetric): 20.0 - 22.0 %
Density (d 20 °C/ 4 °C): 0.875 - 0.880

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.208718.0653871210
grade

synthesis grade

grade

synthesis grade

grade

synthesis grade

grade

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

form

liquid

form

powder

form

powder

form

powder

pH

11 ( in H2O)

pH

13 (20 °C, 5 g/L in H2O)

pH

-

pH

-

storage temp.

15-25°C

storage temp.

-

storage temp.

2-8°C

storage temp.

-

bp

91 °C/1013 hPa

bp

-

bp

-

bp

-


pictograms

FlameCorrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1A

supp_hazards

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

73.4 °F

flash_point_c

23 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters