Skip to Content
Merck

Skip To

8.16014

(R)-(+)-Cysteine

for synthesis

Synonym(s):

(R)-(+)-Cysteine, α-Amino-β-mercapto propionic acid, αlpha-Amino-β-mercapto propionic acid, Cys, αlpha-Amino-beta-mercapto propionic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g
Please contact Customer Service for Availability
CZK 995.00
100 g
Please contact Customer Service for Availability
CZK 3,400.00

About This Item

Empirical Formula (Hill Notation):
C3H7NO2S
CAS Number:
Molecular Weight:
121.16
UNSPSC Code:
12352209
EC Index Number:
200-158-2
NACRES:
NA.22
MDL number:

CZK 995.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

autoignition temp.

420 °C

potency

1890 mg/kg LD50, oral (Rat)

pH

4.5-5.5 (20 °C, 100 g/L in H2O)

mp

220-228 °C

bulk density

300 kg/m3

storage temp.

2-30°C

SMILES string

SCC([N+H3])C(=O)[O-]

InChI

1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)

InChI key

XUJNEKJLAYXESH-UHFFFAOYSA-N

General description

Cysteine is used as a building block in the synthesis of peptides due to its ability to form disulfide bonds. Additionally, it functions as a chiral auxiliary in asymmetric synthesis, facilitating the induction of chirality in the resulting products.

Analysis Note

Assay (HClO₄): ≥ 98.0 %
Spec. rotation [α²0/D (c=8 in HCl 1 mol/l): +8.0 - +9.5 °
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C735295437W326305
form

solid

form

crystalline powder

form

-

form

-

storage temp.

2-30°C

storage temp.

-

storage temp.

2-8°C

storage temp.

-

Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

300

mp

220-228 °C

mp

240 °C (dec.) (lit.)

mp

240 °C (dec.) (lit.)

mp

240 °C (dec.) (lit.)

bulk density

300 kg/m3

bulk density

-

bulk density

-

bulk density

-

autoignition temp.

420 °C

autoignition temp.

-

autoignition temp.

-

autoignition temp.

-


Storage Class

13 - Non Combustible Solids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters