Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
10 g | Please contact Customer Service for Availability | CZK 4,260.00 |
About This Item
Empirical Formula (Hill Notation):
C10H14O2
CAS Number:
Molecular Weight:
166.22
UNSPSC Code:
12352115
EC Index Number:
233-814-1
NACRES:
NA.22
MDL number:
CZK 4,260.00
Please contact Customer Service for Availability
Quality Segment
assay
≥97.0% (HPLC)
mp
197-203 °C (External MSDS)
solubility
slightly soluble 1.7 g/L
storage temp.
2-30°C
SMILES string
O=C1C2(C(C(CC2)C1=O)(C)C)C
InChI
1S/C10H14O2/c1-9(2)6-4-5-10(9,3)8(12)7(6)11/h6H,4-5H2,1-3H3
InChI key
VNQXSTWCDUXYEZ-UHFFFAOYSA-N
Analysis Note
Assay (HPLC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 197 °C
Melting range (upper value): ≤ 200 °C
Identity (IR): passes test
Melting range (lower value): ≥ 197 °C
Melting range (upper value): ≤ 200 °C
Identity (IR): passes test
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.