Skip to Content
Merck

Skip To

8.14364

Didecyldimethylammonium chloride

(50% solution in 2-propanol/water 2:3) for synthesis

Synonym(s):

N-Decyl-N,N-dimethyl-1-decanaminium chloride, DDAC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mL
Please contact Customer Service for Availability
CZK 800.00
1 L
Please contact Customer Service for Availability
CZK 2,670.00

About This Item

NACRES:
NA.22
UNSPSC Code:
12352107
Assay:
50.0 - 55.0%
Form:
liquid

CZK 800.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

50.0 - 55.0%

form

liquid

potency

658 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rat)

reaction suitability

core: ammonium

pH

6.5-8.0 (20 °C, 10 g/L in H2O)

bp

88 °C/1013 hPa

transition temp

flash point 29 °C

density

0.90 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Assay (argentometric): 50.0 - 55.0 %
Density (d 20 °C/ 4 °C): 0.900 - 0.910

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.024488.2039434466
assay

50.0 - 55.0%

assay

-

assay

-

assay

-

reaction suitability

core: ammonium

reaction suitability

-

reaction suitability

-

reaction suitability

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

bp

88 °C/1013 hPa

bp

-

bp

-

bp

-

density

0.90 g/cm3 at 20 °C

density

-

density

-

density

-


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

84.2 °F

flash_point_c

29 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters