Skip to Content
Merck

Skip To

810037

Probumin® Bovine Serum Albumin Universal Grade

pack of 1 kg, ≥98%, USA Origin, Tested for IgG , ELISA: suitable, enzyme immunoassay: suitable, Serology: suitable

Synonym(s):

Albumin, BSA, Bovine Serum, Bovine Serum Albumin, Fraction V

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 kg

Estimated to ship onJuly 15, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 27,500.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41141801
NACRES:
NA.28
Biological source:
bovine serum
Assay:
≥98% (purity)
Form:
lyophilized powder
Technique(s):
ELISA: suitable, enzyme immunoassay: suitable
Impurities:
IgG, tested

CZK 27,500.00


Estimated to ship onJuly 15, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Probumin® Bovine Serum Albumin Universal Grade, pack of 1 kg

biological source

bovine serum

Quality Segment

assay

≥98% (purity)

form

lyophilized powder

shelf life

48 mo.

mol wt

apparent mol wt 66 kDa

purified by

heat shock fractionation

packaging

pack of 1 kg

manufacturer/tradename

Probumin®

origin

USA origin

technique(s)

ELISA: suitable, enzyme immunoassay: suitable

dilution

(Serology)

impurities

IgG, tested

loss

≤5%

pH range

5.0-5.4

anion traces

chloride (Cl-): 0.0-3.0 mg/g

cation traces

Na: 0.0-10.0 mg/g

foreign activity

Bluetongue, none detected, Vesicular Stomatitis, none detected

storage temp.

2-8°C

General description

This BSA powder is manufactured by a proprietary heat-shock fractionation process, using caprylic acid as an albumin stabilizer. A highly consistent and widely used grade of BSA powder for a wide spectrum of immunoassays, cell culture and microbial fermentation applications. This BSA is low in sodium and chloride as well as highly monomeric. Purity specification is >= 98%; typical purity is 100%.

Application

Probumin® BSA products are fit for use as blockers, protein/antibody (conjugate) stabilizers, and in immunoassays (EIA, RIA and serology systems).

Features and Benefits

  • High-quality and reliability. Assurance of consistency in performance and supply.
  • Proprietary heat shock and cold solvent fractionation process optimizes the protein yield, purity, and efficacy of BSA. Large batch capacity.
  • Safe serum sourcing from USDA-licensed facilities exclusively from U.S. cattle.
  • Standard product.
  • Wide range of utility.

Legal Information

Probumin is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Probumin® BSA grades are distinguished by differences in production. The use of terms such as ′Diagnostic Grade′ or ′Vaccine Grade′ in product names solely indicates that said products are commonly used by customers in those applications. We have not qualified every single product in every conceivable application. Users should qualify Probumin® BSA grades using the specific reagents, conditions, and constraints of their assay.
For further manufacturing use only. Not for human or animal consumption.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
810036810667810033
technique(s)

ELISA: suitable, enzyme immunoassay: suitable

technique(s)

blocking: suitable, enzyme immunoassay: suitable

technique(s)

enzyme immunoassay: suitable, protein quantification: suitable

technique(s)

ELISA: suitable, enzyme immunoassay: suitable

origin

USA origin

origin

USA origin

origin

USA origin

origin

USA origin

assay

≥98% (purity)

assay

≥98% (purity)

assay

≥98% (purity)

assay

≥98% (purity)

biological source

bovine serum

biological source

bovine serum

biological source

bovine serum

biological source

bovine serum

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder


Still not finding the right product?


Storage Class

11 - Combustible Solids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.






Global Trade Item Number

SKUGTIN
81003704053252396311

Questions

Reviews

No rating value

Active Filters