Skip to Content
Merck

Skip To

8.09622

Ethyl acetoacetate

for synthesis

Synonym(s):

Ethyl acetoacetate, EAA, Acetoacetic acid ethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 mL
Please contact Customer Service for Availability
CZK 744.00
1 L
Please contact Customer Service for Availability
CZK 1,310.00
2.5 L
Please contact Customer Service for Availability
CZK 2,330.00
25 L
Please contact Customer Service for Availability
CZK 19,000.00

About This Item

Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14
UNSPSC Code:
12352108
EC Index Number:
205-516-1
NACRES:
NA.22
MDL number:
Assay:
≥98.0% (GC)
Form:
liquid

CZK 744.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.26 hPa ( 20 °C)

Quality Segment

assay

≥98.0% (GC)

form

liquid

autoignition temp.

350 °C

potency

3980 mg/kg LD50, oral (Rat), >5000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.0-54 % (v/v)

pH

4.0 (20 °C, 110 g/L in H2O)

mp

-53.3 °C

transition temp

flash point 73.5 °C

solubility

130.3 g/L

density

1.03 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CC)C(=O)CC(=O)C

InChI

1S/C6H10O3/c1-3-9-6(8)4-5(2)7/h3-4H2,1-2H3

InChI key

XYIBRDXRRQCHLP-UHFFFAOYSA-N

Application

Ethyl acetoacetate can be used as a reactant to synthesize:
  • 7-hydroxycoumarin derivatives via Pechmann condensation reaction with 1,3-dihydroxybenzene in the presence of acid catalysts.
  • 2,6-disubstituted piperidine alkaloid, (−)-pinidinone via stereoselective α-aminoallylation followed by Grubbs′ olefin cross-metathesis reaction.
  • Michael addition products via Michael addition reaction with chalcones and azachalcones in the presence of a base catalyst.
  • α, β-unsaturated carbonyl compounds via Knoevenagel condensation reaction with glyceraldehyde acetonide in the presence of a base catalyst.

It can be also utilized as a reactant in the transesterification and asymmetric hydrogenation reactions to produce valuable products.

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.028 - 1.030
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
688983W24150400410
assay

≥98.0% (GC)

assay

≥99.0% (GC)

assay

≥99%

assay

≥99.0% (GC)

solubility

130.3 g/L

solubility

water: soluble 130 g/L at 20 °C

solubility

water: soluble 130 g/L at 20 °C

solubility

water: soluble 130 g/L at 20 °C

Quality Level

200

Quality Level

200

Quality Level

300, 400

Quality Level

200

form

liquid

form

liquid

form

-

form

-

mp

-53.3 °C

mp

−43 °C (lit.)

mp

−43 °C (lit.)

mp

−43 °C (lit.)

density

1.03 g/cm3 at 20 °C

density

1.029 g/mL at 20 °C (lit.)

density

1.029 g/mL at 20 °C (lit.)

density

1.029 g/mL at 20 °C (lit.)


Still not finding the right product?

Explore all of our products under Ethyl acetoacetate


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

164.3 °F - closed cup

flash_point_c

73.5 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Synthesis of 7-hydroxycoumarins catalysed by solid acid catalysts
Hoefnagel AJ, et al.
Journal of the Chemical Society. Chemical Communications, (2), 225-226 (1995)
Condensation of glyceraldehyde acetonide with ethyl acetoacetate over Mg, Al-mixed oxides derived from hydrotalcites
Veloso Claudia O, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 107(1-2), 23-30 (2008)
An Expeditious Stereoselective Synthesis of (-)-Pinidinone from Ethyl Acetoacetate
Damodar K and Jun Jong-Gab
Bulletin of the Korean Chemical Society,, 37(4), 571-575 (2016)



Global Trade Item Number

SKUGTIN
809622902504022536402109
809622250004022536402086
809622010004022536402062
809622100004022536402079

Questions

Reviews

No rating value

Active Filters