Skip to Content
Merck

Skip To

8.03240

N-(2-Chloropropyl)-N,N-dimethylammonium chloride

for synthesis

Synonym(s):

N-(2-Chloropropyl)-N,N-dimethylammonium chloride, 2-Chloro-1-dimethylaminopropane hydrochloride, 2-DMPC, 2-Chloro-N,N-dimethylpropylamine hydrochloride, 1-Dimethylaminoisopropyl chloride hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
250 g
Please contact Customer Service for Availability
CZK 2,450.00
CZK 1,837.50

About This Item

Linear Formula:
(CH3CHClCH2)(CH3)2N · HCl
CAS Number:
Molecular Weight:
158.07
UNSPSC Code:
12352107
EC Index Number:
224-971-7
NACRES:
NA.22
MDL number:
Form:
powder
Solubility:
2000 g/L

CZK 1,837.50

List PriceCZK 2,450.00Save 25%

Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

potency

386 mg/kg LD50, oral (Rat)

pH

5-6 (20 °C, 500 g/L in H2O)

mp

188-191 °C

transition temp

flash point 220 °C

solubility

2000 g/L

bulk density

350 kg/m3

storage temp.

2-30°C

SMILES string

[Cl-].ClC(C[N+H](C)C)C

InChI

1S/C5H12ClN.ClH/c1-5(6)4-7(2)3;/h5H,4H2,1-3H3;1H

InChI key

OCWGRWAYARCRTQ-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.024488.20394D141208
form

powder

form

powder

form

powder

form

crystals

solubility

2000 g/L

solubility

2000 g/L

solubility

-

solubility

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

mp

188-191 °C

mp

205-208 °C

mp

133-136 °C

mp

201-204 °C (lit.)

pH

5-6 (20 °C, 500 g/L in H2O)

pH

-

pH

-

pH

-

potency

386 mg/kg LD50, oral (Rat)

potency

238 mg/kg LD50, oral (Rat), 290 mg/kg LD50, skin (Rabbit)

potency

-

potency

-


pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
803240025004022536379692

Questions

Reviews

No rating value

Active Filters