Skip to Content
Merck

Skip To

8.02260

Bromoacetic acid

for synthesis

Synonym(s):

Bromoacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
5 g
Please contact Customer Service for Availability
CZK 668.00
250 g
Please contact Customer Service for Availability
CZK 1,370.00
1 kg
Please contact Customer Service for Availability
CZK 3,750.00

About This Item

Empirical Formula (Hill Notation):
C2H3BrO2
CAS Number:
Molecular Weight:
138.95
UNSPSC Code:
12352106
EC Index Number:
201-175-8
NACRES:
NA.22
MDL number:
Assay:
≥98.0% (GC)
Form:
crystals

CZK 668.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.158 hPa ( 25 °C)

Quality Level

assay

≥98.0% (GC)

form

crystals

potency

50 mg/kg LD50, oral (Rat)

bp

208 °C/1013 hPa

mp

46-49 °C

transition temp

flash point >112 °C

density

1.93 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

BrCC(=O)O

InChI

1S/C2H3BrO2/c3-1-2(4)5/h1H2,(H,4,5)

InChI key

KDPAWGWELVVRCH-UHFFFAOYSA-N

Application


  • Bromoacetic acid induces neurogenic injury in the chicken brain: This research investigates the neurotoxic effects of Bromoacetic acid, demonstrating its impact on oxidative stress and inflammatory pathways in avian species (Liu et al., 2022).

  • Delayed local chemical burns following dermal exposure to bromoacetic acid: A case study documenting the severe dermatological effects following exposure to bromoacetic acid, providing crucial insights into its hazardous nature (Yeung et al., 2024).

  • Toxicity Effect of Bromoacetic Acid on MCF7 Breast Cancer Cell Line: This study explores the cytotoxic effects of Bromoacetic Acid on cancer cells, with an emphasis on the mechanisms of apoptosis induced by this compound (Bahman et al., 2020).


Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.01636B56307901610
assay

≥98.0% (GC)

assay

≥98.0% (GC)

assay

97%

assay

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

-

mp

46-49 °C

mp

-38 °C

mp

47-49 °C (lit.)

mp

-

form

crystals

form

liquid

form

crystals

form

liquid

density

1.93 g/cm3 at 20 °C

density

1.50 g/cm3 at 20 °C

density

-

density

1.0264 g/mL

bp

208 °C/1013 hPa

bp

158-161 °C/1013 hPa

bp

208 °C (lit.)

bp

-


Still not finding the right product?

Explore all of our products under Bromoacetic acid


signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1A - Skin Sens. 1

Storage Class

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
802260025004022536372440
802260100004022536372457
802260000504022536372433

Questions

Reviews

No rating value

Active Filters