Skip to Content
Merck

Skip To

8.00888

Hexacarbonyltungsten

for synthesis

Synonym(s):

Hexacarbonyltungsten, Tungsten carbonyl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 g
Please contact Customer Service for Availability
CZK 9,000.00
25 g
Please contact Customer Service for Availability
CZK 17,200.00

About This Item

Linear Formula:
W(CO)6
CAS Number:
Molecular Weight:
351.90
UNSPSC Code:
12161603
EC Index Number:
237-880-2
NACRES:
NA.22
MDL number:

CZK 9,000.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

1.4 hPa ( 67 °C)

Quality Segment

form

powder

mp

169-171 °C

density

2.65 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[W].O=[C].O=[C].O=[C].O=[C].O=[C].O=[C]

InChI

1S/6CO.W/c6*1-2;

InChI key

FQNHWXHRAUXLFU-UHFFFAOYSA-N

Analysis Note

Assay (ex W): ≥ 98.0 %
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
241431755737808822
form

powder

form

solid

form

solid

form

crystals

storage temp.

2-30°C

storage temp.

-

storage temp.

-

storage temp.

-

mp

169-171 °C

mp

150 °C (lit.)

mp

150 °C (lit.)

mp

-

density

2.65 g/cm3 at 20 °C

density

2.65 g/mL at 25 °C (lit.)

density

2.65 g/mL at 25 °C (lit.)

density

-

vapor pressure

1.4 hPa ( 67 °C)

vapor pressure

1.2 mmHg ( 67 °C)

vapor pressure

1.2 mmHg ( 67 °C)

vapor pressure

-

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

100


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters