Skip to Content
Merck

Skip To

8.00579

Ethyl methacrylate

(stabilised) for synthesis

Synonym(s):

Ethyl methacrylate, Ethyl 2-methylpropenoate, Methacrylic acid ethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 mL
Please contact Customer Service for Availability
CZK 668.00
500 mL
Please contact Customer Service for Availability
CZK 1,250.00
CZK 937.50

About This Item

Linear Formula:
CH2=C(CH3)COOC2H5
CAS Number:
Molecular Weight:
114.14
UNSPSC Code:
12352108
EC Index Number:
202-597-5
NACRES:
NA.22
MDL number:

CZK 668.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

21.3 hPa ( 20 °C)

Quality Segment

assay

≥99.0% (GC)

form

liquid

autoignition temp.

329 °C

potency

14600 mg/kg LD50, oral (Rat), >9130 mg/kg LD50, skin (Rabbit)

expl. lim.

1.8 % (v/v)

bp

119-121 °C/1013 hPa

mp

-61--59 °C

transition temp

flash point 18 °C

solubility

5.1 g/L

density

0.913 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CC)C(=O)C(=C)C

InChI

1S/C6H10O2/c1-4-8-6(7)5(2)3/h2,4H2,1,3H3

InChI key

SUPCQIBBMFXVTL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.912 - 0.913
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.005908.005888.00585
assay

≥99.0% (GC)

assay

≥99.0% (GC)

assay

≥97.0% (GC)

assay

≥99.0% (GC)

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

density

0.913 g/cm3 at 20 °C

density

0.94 g/cm3 at 20 °C

density

1.07 g/cm3 at 20 °C

density

0.89 g/cm3 at 20 °C

solubility

5.1 g/L

solubility

-

solubility

-

solubility

-

mp

-61--59 °C

mp

-48 °C

mp

-60 °C

mp

-75 °C

form

liquid

form

liquid

form

liquid

form

liquid


Still not finding the right product?

Explore all of our products under Ethyl methacrylate


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

66.2 °F - closed cup

flash_point_c

19 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
800579010004022536360324
800579050004022536360331

Questions

Reviews

No rating value

Active Filters