Skip to Content
Merck

Skip To

8.00423

3-Amino-1-propanol

for synthesis

Synonym(s):

3-Amino-1-propanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mL
Please contact Customer Service for Availability
CZK 667.00
250 mL
Please contact Customer Service for Availability
CZK 1,120.00

About This Item

Empirical Formula (Hill Notation):
C3H9NO
CAS Number:
Molecular Weight:
75.11
UNSPSC Code:
12352104
EC Index Number:
205-864-4
NACRES:
NA.22
MDL number:
Assay:
≥99.0% (GC)
Form:
liquid

CZK 667.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.149 hPa ( 25 °C)

Quality Segment

assay

≥99.0% (GC)

form

liquid

autoignition temp.

385 °C

potency

1300 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rat)

expl. lim.

2.5-10.6 % (v/v)

pH

11.6 (20 °C, 10 g/L in H2O)

kinematic viscosity

29.9 cSt(23 °C)

bp

187-190 °C

mp

11.5-12.4 °C

transition temp

flash point 100 °C

density

0.99 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

NCCCO

InChI

1S/C3H9NO/c4-2-1-3-5/h5H,1-4H2

InChI key

WUGQZFFCHPXWKQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.986 - 0.988
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.014658.140998.14513
assay

≥99.0% (GC)

assay

93.0-100.0% (GC)

assay

-

assay

≥97.0% (GC)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

density

0.99 g/cm3 at 20 °C

density

0.95 g/cm3 at 25 °C

density

-

density

1.16 g/cm3 at 20 °C

mp

11.5-12.4 °C

mp

30-31 °C

mp

204 °C (External MSDS)

mp

-

bp

187-190 °C

bp

165 °C/1013 hPa (anhydrous)

bp

-

bp

-

form

liquid

form

solid

form

powder

form

liquid


Still not finding the right product?

Explore all of our products under 3-Amino-1-propanol


pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 1

flash_point_f

212.0 °F - Pensky-Martens closed cup

flash_point_c

100 °C - Pensky-Martens closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters