Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 100 mL | Estimated to ship onJune 03, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 1,990.00 |
About This Item
NACRES:
NA.77
UNSPSC Code:
41106500
CZK 1,990.00
Estimated to ship onJune 03, 2026Details
form
liquid
Quality Segment
manufacturer/tradename
Novagen®
storage condition
OK to freeze
storage temp.
2-8°C
General description
Pretreatment of fixed tissues with an antigen retrieval buffer can dramatically improve antibody staining. For tissues and antibodies for which antigen retrieval is necessary, Novagen is pleased to introduce the AntigenPlus Buffer Set designed for antigen retrieval applications in fixed, paraffin embedded tissues. The Buffer Set consists of three different 5X AntigenPlus Buffers (pH 6, pH 7.4 and pH 10) that can be tested with the tissues of interest.
Each buffer is also available individually in 100 ml size:
5X AntigenPlus Buffer (pH 6)
5X AntigenPlus Buffer (pH 7.4)
5X AntigenPlus Buffer (pH 10)
Each buffer is also available individually in 100 ml size:
5X AntigenPlus Buffer (pH 6)
5X AntigenPlus Buffer (pH 7.4)
5X AntigenPlus Buffer (pH 10)
Legal Information
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level - |
| form liquid | form liquid | form liquid | form liquid |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. - | storage temp. - |
| storage condition OK to freeze | storage condition OK to freeze | storage condition - | storage condition - |
| manufacturer/tradename Novagen® | manufacturer/tradename Novagen® | manufacturer/tradename Upstate® | manufacturer/tradename Chemicon®, IHC Select |
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 71289-3 | 04055977253979 |



