Skip to Content
Merck

Skip To

69968-M

Biotinylated T7 Tag Monoclonal Antibody

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.41
UNSPSC Code:
51111800
Clone:
-
Species reactivity:
-
Application:
Citations:
-




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library