Skip to Content
Merck

Skip To

69265

Goat Anti-Rabbit IgG AP Conjugate (Fc specific)

Novagen®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
40 μL

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 3,030.00

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Clone:
-
Species reactivity:
-
Application:
Citations:
-

CZK 3,030.00


Available to ship TODAYDetails


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


antibody product type

secondary antibodies

Quality Level

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
69266401414401445
shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

manufacturer/tradename

Novagen®

manufacturer/tradename

Novagen®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

storage condition

OK to freeze, avoid repeated freeze/thaw cycles, protect from light

storage condition

do not freeze


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
69265-304055977259346

Questions

Reviews

No rating value

Active Filters