Skip to Content
Merck

Skip To

69238

Rat Genomic DNA

High molecular weight, high purity DNA for any application

Synonym(s):

Rat DNA, Rat genomic DNA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg

Estimated to ship onApril 28, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 7,980.00

About This Item

NACRES:
NA.51
UNSPSC Code:
12352208
Form:
liquid
Solubility:
water: soluble at 20 °C
Storage temp.:
2-8°C

CZK 7,980.00


Estimated to ship onApril 28, 2026Details


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

solubility

water: soluble at 20 °C

shipped in

wet ice

storage temp.

2-8°C

General description

Novagen genomic DNA is a purified preparation of high molecular weight genomic DNA obtained from various organisms. During the isolation process, measures are taken to minimize shearing and eliminate contaminants that could disrupt enzymatic procedures. It has been qualified for genomic analysis, including library construction and PCR, as at least 90% of the DNA is greater than 100 kbp in size, with contour-clamped homogeneous electric field (CHEF) electrophoresis analysis showing sizes greater than 50 kbp for S. cerevisiae. Notably, no inhibition of restriction enzyme activity has been observed, and the DNA is free from protein and RNA contaminants.

Application

Ratgenomic DNA has been used as a template for the amplification of theprogression elevated gene-3 (PEG-3) gene using polymerase chain reaction (PCR).

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
700436923970040
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

manufacturer/tradename

Novagen®

manufacturer/tradename

Novagen®

manufacturer/tradename

Novagen®

manufacturer/tradename

Novagen®

form

liquid

form

liquid

form

liquid

form

liquid

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

solubility

water: soluble at 20 °C

solubility

-

solubility

water: soluble at 20 °C

solubility

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
69238-304055977274639

Questions

  1. Regarding cat no. 69238 Rat Genomic DNA, which extraction method was used to extract the HMW DNA? We are specifically interested in rat mitochondrial DNA, is this still included?

    1 answer
    1. The isolation/extraction method for Rat Genomic DNA is proprietary and unavailable. The DNA is obtained from multiple animals from a single strain (Sprague Dawley) and a mixed sex population. The isolation protocol minimizes shearing and removes contaminants that inhibit enzymatic procedures. The DNA's origin, whether a mixture of mitochondrial and nuclear, solely mitochondrial, or nuclear, is not evaluated as part of the quality control process.

      Helpful?

Reviews

No rating value

Active Filters