Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Linear Formula:
(O2N)3C6H2OH
CAS Number:
Molecular Weight:
229.10
UNSPSC Code:
12352106
Beilstein/REAXYS Number:
423400
Colour Index Number:
10305
MDL number:
Grade:
purified grade
Pricing and availability is not currently available.
grade
purified grade
Quality Segment
expl. lim.
0.01 %
mp
122-123 °C (dried material) (lit.)
SMILES string
Oc1c(cc(cc1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O
InChI
1S/C6H3N3O7/c10-6-4(8(13)14)1-3(7(11)12)2-5(6)9(15)16/h1-2,10H
InChI key
OXNIZHLAWKMVMX-UHFFFAOYSA-N
Still not finding the right product?
Explore all of our products under Picric acid
signalword
Danger
Storage Class
4.1A - Other explosive hazardous materials
flash_point_f
302.0 °F
flash_point_c
150 °C
hcodes
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Expl. 1.1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

