Skip to Content
Merck

Skip To

566321

Sirtinol

InSolution, ≥97%, inhibitor of sirtuin class of histone deacetylase (HDAC) activity

Synonym(s):

InSolution Sirtinol, 2-[(2-Hydroxynaphthalen-1-ylmethylene)amino]-N-(1-phenethyl)benzamide, Sir Two Inhibitor Naphthol, SIRT1 Inhibitor I, SIRT1/2 Inhibitor I, SIRT2 Inhibitor III

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C26H22N2O2
Molecular Weight:
394.47
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥97% (HPLC)
Form:
liquid
Quality level:
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light


Quality Level

assay

≥97% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

shipped in

wet ice

storage temp.

2-8°C

General description

A cell-permeable 2-hydroxy-1-napthaldehyde derivative that acts as a specific and a direct inhibitor of sirtuin class of histone deacetylase (HDAC) activity. Does not affect human HDAC1. Reported to block Sir2p transcriptional silencing activity in vivo (IC50 = 25 µM) and NAD-dependent HDAC activity in purified recombinant yeast Sir2p and human SIRT2 in vitro (IC50 = 68 µM and 38 µM, respectively).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ySir2
Product does not compete with ATP.
Reversible: no
Target IC50: 48 µM, 131 µM and 58 µM for ySir2, hSIRT1 and hSIRT2, respectively

Packaging

Packaged under inert gas

Physical form

A 10 mM (1 mg/254 µl) solution of Sirtinol (Cat. No. 566320) in DMSO.

Preparation Note

Following initial thaw, aliquot and refrigerate (4°C).

Other Notes

Mai, A., et al. 2005. J. Med. Chem.48, 7789.
Grozinger, C.M., et al. 2001. J. Biol. Chem.276, 38837.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
566320566323569396
assay

≥97% (HPLC)

assay

≥97% (HPLC)

assay

≥95% (HPLC)

assay

≥97% (HPLC)

form

liquid

form

solid

form

solid

form

liquid

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−70°C

storage condition

OK to freeze, protect from light, desiccated (hygroscopic)

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

188.6 °F - closed cup - (Dimethylsulfoxide)

flash_point_c

87 °C - closed cup - (Dimethylsulfoxide)



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

"The SIRTainty™ Class III HDAC Assay utilizes a novel patent pending technology for the sensitive detection of all known sirtuin family members. Unlike conventional assays that are dependent upon a single pre-labeled fluorescently tagged substrate, the SIRTainty Class III HDAC Assay employs untagged acetylated peptide substrates. This approach not only enables unparalleled flexibility in your choice of sirtuin isoform and peptide substrate, but also eliminates the potential for artifacts due to the use of artificial substrates containing bulky fluorophores."


Yi-Fan Lin et al.
iScience, 24(10), 103118-103118 (2021-10-09)
The mitochondrial unfolded protein response (UPRmt) is an organellar stress signaling pathway that functions to detect and restore disruption of mitochondrial proteostasis. The UPRmt is involved in a wide range of physiological and disease conditions, including aging, stem cell maintenance



Global Trade Item Number

SKUGTIN
566321-1MG04055977191387

Questions

Reviews

No rating value

Active Filters