Skip to Content
Merck

Skip To

565835

3AC

≥98% (1H-NMR), solid, SHIP1 inhibitor, Calbiochem®

Synonym(s):

SHIP1 Inhibitor, 3AC, 3α-Aminocholestane, SH2-domain-containing inositol 5ʹ-phosphatase 1 (SHIP1) Inhibitor, 3AC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
10 mg

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 4,990.00

About This Item

Empirical Formula (Hill Notation):
C27H49N
Molecular Weight:
387.68
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (1H-NMR)
Form:
solid
Quality level:
Storage condition:
OK to freeze

CZK 4,990.00


Available to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

SHIP1 Inhibitor, 3AC, The SHIP1 Inhibitor, 3AC controls the biological activity of SHIP1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Quality Level

assay

≥98% (1H-NMR)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

ethanol: 100 mg/mL, DMSO: insoluble

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable steroidal compound that selectively inhibits SHIP1-, but not SHIP2-, catalyzed PI(3,4,5)P3-to-PI(3,4)P2 dephosphorylation (IC50 = 10 µM vs. >1 mM, respectively), while exhibiting little activity against the PTEN-catalyzed PI(3,4,5)P3-to-PI(4,5)P2 conversion (IC50 >1 mM). 3AC in vivo treatment (2.292 umol/200 µl/mouse/daily i.p. for 7 d) is reported to boost both peripheral blood granulocyte production (by 4- to 5-fold) and the GVHD- (graft-versus-host disease) suppressing Mac1+Gr1+ MIR (myeloid immunoregulatory) cell population in lymphoid tissues (by 5.55- and 11.53-fold in spleen and lymph nodes, respectively), resulting in an impaired splenocyte allogeneic T cell response in MLR (mixed-leukocyte/lymphocyte reaction) assays (by ≥95%). Similarly, 3AC-treated (9.4 µM; 24 h) primary human PBMCs greatly lose their ability in priming allogeneic T cell response in MLR assays (by 74.5%). In addition, a faster blood cell recovery is also observed in mice receiving 3AC (2.292 umol/200 µl/mouse/daily i.p. for 7 d) after sub-lethal irradiation (550 Rads). Selectively inhibits the growth and survival of SHIP1-dependent human KG-1 and murine C1498 myelogenous leukemia lines (IC50 = 11.5 µM), but not the SHIP1-negative human CML K562 and osteosarcoma MG63 lines. SHIP2 Inhibitor, AS1938909 is also available (Cat. No. 565840).

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Brooks, R., et al. 2010. J. Immunol.184, 3582.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
565840124009407625
form

solid

form

solid

form

solid

form

solid

assay

≥98% (1H-NMR)

assay

≥95% (HPLC)

assay

≥98% (NMR)

assay

≥98% (HPLC)

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

solubility

ethanol: 100 mg/mL, DMSO: insoluble

solubility

DMSO: 50 mg/mL

solubility

DMSO: 10 mg/mL

solubility

DMSO: 50 mg/mL, light pink-purple


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Wendy M Kandell et al.
PloS one, 15(2), e0225820-e0225820 (2020-02-11)
NK cell migration and activation are crucial elements of tumor immune surveillance. In mammary carcinomas, the number and function of NK cells is diminished, despite being positively associated with clinical outcome. MicroRNA-155 (miR-155) has been shown to be an important



Global Trade Item Number

SKUGTIN
565835-10MG04055977191929

Questions

Reviews

No rating value

Active Filters