Skip to Content
Merck

Skip To

539251

Protein A, Alkaline Phosphatase Conjugate

This Protein A, Alkaline Phosphatase Conjugate is validated for use in Direct Immunoassay.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg

Estimated to ship onJune 02, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 6,790.00

About This Item

NACRES:
NA.41
UNSPSC Code:
41116133
Form:
liquid
Biological source:
Staphylococcus aureus

CZK 6,790.00


Estimated to ship onJune 02, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Staphylococcus aureus

Quality Segment

form

liquid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

dilution

(Direct Immunoassay (1:5000))

shipped in

wet ice

storage temp.

2-8°C

General description

Native protein A conjugated to alkaline phosphatase. Can be used instead of an alkaline phosphatase-conjugated secondary antibody in many immunological assays.
Native protein A from Staphylococcus aureus conjugated to alkaline phosphatase.

Application

Direct Immunoassay (1:5000)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In 50 mM Tris, 1 mM MgCl₂, 1% BSA, pH 8.0.

Analysis Note

Single band by SDS-PAGE

Other Notes

p-Nitrophenyl phosphate, 1 mg/ml in 10% diethanolamine, pH 9.8, 1 mM MgCl2 at 25°C, was used as substrate for testing immunoenzyme reactivity. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
539253539305539322
biological source

Staphylococcus aureus

biological source

Staphylococcus aureus

biological source

(Streptococcus species)

biological source

(Streptococcus species)

form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage condition

do not freeze

storage condition

do not freeze

storage condition

do not freeze

storage condition

do not freeze

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters