Skip to Content
Merck
All Photos(2)

Key Documents

5.32912

Sigma-Aldrich

CIP2A Inhibitor, TD-19

Synonym(s):

CIP2A Inhibitor, TD-19, N4-(3-ethynylphenyl)-6,7-dimethoxy-N2-(4-phenoxyphenyl)quinazoline-2,4-diamine, TD19

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H24N4O3
CAS Number:
Molecular Weight:
488.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL (with warming; solution will fall out of solution at room temperature)
DMF: soluble

storage temp.

−20°C

General description

A cell-permeable chemically modified derivative of erlotinib that is shown to be more effective in inducing apoptosis in H358, H441, H460, and A549 lung cancer cell lines (~10 µM). Highly effective in reducing the viability and inducing apotosis in erlotinib resistantH460 cells over-expressing CIP2A (cancerous inhibitor of protein phosphatase 2A ) that are resistant to erlotinib. Exhibits low binding affinity to the ATP binding site of the EGFR tyrosine kinase domain and has minimal effect on the phosphorylation of EGFR. Its anti-tumor effects are shown to be mediated via down-regulation of CIP2A and p-AKT expression and increased PP2A activity. Suppresses the growth of H460 xenograft tumors in NCr nude mice by approximately 80% (10 mg/kg. p.o. daily).
A cell-permeable chemically modified derivative of erlotinib that is shown to be more effective in inducing apoptosis in H358, H441, H460, and A549 lung cancer cell lines (~10 µM). Highly effective in reducing the viability and inducing apotosis in erlotinib resistantH460 cells over-expressing CIP2A (cancerous inhibitor of protein phosphatase 2A ) that are resistant to erlotinib. Exhibits low binding affinity to the ATP binding site of the EGFR tyrosine kinase domain and has minimal effect on the phosphorylation of EGFR. Its anti-tumor effects are shown to be mediated via down-regulation of CIP2A and p-AKT expression and increased PP2A activity. Suppresses the growth of H460 xenograft tumors in NCr nude mice by approximately 80% (10 mg/kg. p.o. daily).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Primary Target
CIP2A

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Chao, T.T., et al. 2014. J. Pharm. Exp. Ther.351,352.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service