Skip to Content
Merck

Skip To

5.32306

PAS Kinase Inhibitor, BioE-1115

Synonym(s):

PAS Kinase Inhibitor, BioE-1115, PASK Inhibitor, 2-(4-Fluorophenyl)-3-(isopropyl(methyl)amino)quinoxaline-6-carboxylic acid, BioE1115, Per-Arnt-Sim Domain Kinase Inhibitor, PAS Domain Kinase Inhibitor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H18FN3O2
CAS Number:
Molecular Weight:
339.36
MDL number:
NACRES:
NA.77
UNSPSC Code:
51111800
Assay:
≥97% (HPLC)
Form:
powder
Storage condition:
OK to freeze, protect from light
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥97% (HPLC)

Quality Segment

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable quinoxaline-carboxylic acid compound that acts as a selective PAS kinase/PASK inhibitor (IC50 ~ 4 nM) with little or no potency against a panel of 49 other kinases (IC50 ≥ 10 µM) and effectively inhibits cellular PASK-T307 autophosphorylation (IC50 ~ 1 µM; 16 h drug treatment in PASK-transfected HEK293T cultures with 1% FBS). Similar to siRNA-mediated PASK knockdown, BioE-1115 treatment is shown to effectively prevent sterol regulatory element binding protein SREBP-1c maturation without affecting Akt/mTOR pathway signaling, resulting in impaired cellular SREBP transcription activity in HepG2 cultures (% inhibition/[drug] = 40%/30 µM & & 65%/50 µM by SRE-Luc reporter assay; overnight drug treatment prior to 100 nM insulin stimulation for 6 h). Oral administration is reported to effectively reduce high-frucose diet/HFrD-induced dyslipidemia (% reduction of liver triglyceride/serum triacylglyerol/dose = 48/26/30 mg kg-1 & 63/55/100 mg kg-1; Daily oral dosage administered in the last wk of a 3 wk HFrD period, followed by a 24 h fasting and a 12 refed period prior to tissue collection) and insulin resistance (% reduction of serum glucose/insulin/dose = 23/14/30 mg kg-1 & 28/31/100 mg kg-1) in rats by selectively suppressing SREBP-1 maturation and thereby inhibiting SREBP-1c, but not SRREBP-2, target genes transcription in liver, but not in abdominal fat or gastrocnemius muscle in vivo (% Gpat1/Fasn/Scd1/Acc1/Fae mRNA reduction/plasma [BioE-1115] in µg/mL/dose = 40/34/36/27/34/2.07/10 mg kg-1, 59/56/51/48/54/7.65/30 mg kg-1, 76/5967/62/74/42.2/100 mg kg-1) without affecting liver or body weight.
A cell-permeable, orally available, non-toxic quinoxaline-carboxylic acid based compound that acts as a highly potent, selective, and reversible inhibitor of Per-Arnt-Sim Kinase (PASK; IC50 ~ 4 nM). Exhibits excellent selectivity over 49 other kinases (IC50 >10 µM) and displays about 2,500-fold greater potency for PASK over casein kinase 2α. Blocks PASK autophosphorylation at Thr307 in a dose-dependent manner (IC50 ~ 1 µM) without affecting the insulin-induced phosphorylation of either Akt or S6K. Effectively blocks the maturation of SREBP-1 in hepatic tissue of high fructose fed wild-type Sprague-Dawley rats. Shown to normalize hepatic and serum triglyceride levels, reduce blood glucose levels, and partially reverse insulin resistance in animal models (30 mg/kg. p.o.).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
BioE-1115, PASKi

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PASK

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wu, X., et al. 2014. Cell Rep.8, 242.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
532306000104055977287196

Questions

Reviews

No rating value

Active Filters