Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C17H15N3O3
CAS Number:
Molecular Weight:
309.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
solid
Quality level:
Storage condition:
OK to freeze, protect from light
Product 5.30655 is not currently sold in your country. Contact Technical Service
Product Name
Mitochondrial GPDH Inhibitor, iGP-1,
assay
≥98% (HPLC)
Quality Level
form
solid
potency
6.3 μM IC50
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, protect from light
color
beige
solubility
DMSO: 50 mg/mL
storage temp.
2-8°C
SMILES string
[nH]1c2c(nc1c3ccc(cc3)NC(=O)CCC(=O)O)cccc2
InChI key
ARVPDCQNTCJVSU-UHFFFAOYSA-N
General description
A cell-permeable thiadiazolyl-butyl-pyridazinyl compound that displays increased fluorescence with acidified pH (by 8-fold from pH 7.0 to 1.5; Ex 342 nm & Em 378 nm) and selectively inhibits mitochondrial sn-glycerol 3-phosphate (GP) dehydrogenase (mGPDH) activity (IC50 = 6.3 µM by DCPIP assay using rat skeletal muscle mitochondria preparation with 1 µ Ca2+), but not cytosolic GPDH (up to 80 µM using rabbit cGPDH), by targeting free mGPDH in a GP-competitive manner (KicKiu2-dependent complex I activity and little potency toward unbiquinol/QH2-dependent complex III activity, nor FMN-dependent complex I or II activity. Under conditions where Lactate dehydrogenase-mediated NAD+ generation is blocked by Oxamate in murine cortical synaptosome preparations, a small but significant dependence on mGPDH-mediated GP shuttle for NAD+ generation to support the high glycolytic demand for pyruvate generation from glucose (15 mM) is reported, as evidenced by a more pronounced mitochondrial respiration inhibition in the presence of FCCP and Oligomycin (4 µg/mL; Cat. No. 495455 ) with the combined treatment of 0.5 mM aminooxyacetate (AOA) and 100 µM iGP-1 than with AOA alone.
Biochem/physiol Actions
Cell permeable: yes
Primary Target
mGPDH
mGPDH
Packaging
Packaged under inert gas
Preparation Note
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Other Notes
Orr, A.L., et al. 2014. PLoS One9, e89938.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 5306550001 | 04055977260427 |