Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C26H29ClN2O4S · xH3PO4
Molecular Weight:
501.04
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
liquid
Storage condition:
OK to freeze, avoid repeated freeze/thaw cycles, protect from light
Pricing and availability is not currently available.
Quality Segment
assay
≥95% (HPLC)
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, avoid repeated freeze/thaw cycles, protect from light
storage temp.
−70°C
General description
A water-soluble form of the CaM kinase II inhibitor KN-93 (Cat. No. 422708). Inhibits glucose-induced and forskolin-stimulated insulin release from isolated pancreatic islets of Langerhans. Has no significant effect on protein kinase A activity. Induces G1 cell cycle arrest and apoptosis in NIH 3T3 cells.
Biochem/physiol Actions
Cell permeable: yes
Primary Target
CaM kinase II
CaM kinase II
Reversible: yes
Packaging
Packaged under inert gas
Physical form
A 5 mM (500 µg/167 µL) sterile-filtered solution of KN-93 Phosphate (Cat. No. 422711) in H₂O.
Preparation Note
Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 6 months at -20°C.
Other Notes
Tombes, R.M., et al. 1995. Cell Growth Differ.6, 1063.
Chen, C., et al. 1994. Science266, 291.
Mamiya, N., et al. 1993. Biochem. Biophys. Res. Commun.195, 608.
Nicki, I., et al. 1993. Biochem. Biophys. Res. Commun.191, 255.
Sumi, M., et al. 1991. Biochem. Biophys. Res. Commun.181, 968.
Tokumitsu, H., et al. 1990. J. Biol. Chem.265, 4315.
Chen, C., et al. 1994. Science266, 291.
Mamiya, N., et al. 1993. Biochem. Biophys. Res. Commun.195, 608.
Nicki, I., et al. 1993. Biochem. Biophys. Res. Commun.191, 255.
Sumi, M., et al. 1991. Biochem. Biophys. Res. Commun.181, 968.
Tokumitsu, H., et al. 1990. J. Biol. Chem.265, 4315.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 5083150001 | 04055977261837 |