Skip to Content
Merck
All Photos(1)

Key Documents

487960

Sigma-Aldrich

Nociceptin

Endogenous ligand for the opioid-like G-protein coupled receptor, ORL1 (Opioid Receptor-Like 1).

Synonym(s):

Nociceptin, Orphanin FQ, OFQ, NC, FGGFTGARKSARKLANQ

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CZK 4,670.00

CZK 4,670.00


Estimated to ship onApril 29, 2025Details


Request a Bulk Order

Select a Size

Change View
1 MG
CZK 4,670.00

About This Item

Empirical Formula (Hill Notation):
C79H129N27O22
CAS Number:
Molecular Weight:
1809.04
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

CZK 4,670.00


Estimated to ship onApril 29, 2025Details


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N([C@@H]([C@H](O)C)C(=O)NCC(=O)N[C@@H](C)C(=O)N[C@@H](CCCN\C(=N\[H])\N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CO)C(=O)N[C@@H](C)C(=O)N[C@@H](CCCN\C(=N\[H])\N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(=O)N)C(=O)N[C@@H](CCC(=O)N)C(=O)O)

InChI

1S/C79H129N27O22/c1-41(2)33-54(72(122)95-44(5)66(116)103-56(36-59(84)110)73(123)102-53(77(127)128)27-28-58(83)109)104-70(120)49(23-13-15-29-80)100-69(119)52(26-18-32-90-79(87)88)99-65(115)43(4)96-75(125)57(40-107)105-71(121)50(24-14-16-30-81)101-68(118)51(25-17-31-89-78(85)86)98-64(114)42(3)94-61(112)39-93-76(126)63(45(6)108)106-74(124)55(35-47-21-11-8-12-22-47)97-62(113)38-91-60(111)37-92-67(117)48(82)34-46-19-9-7-10-20-46/h7-12,19-22,41-45,48-57,63,107-108H,13-18,23-40,80-82H2,1-6H3,(H2,83,109)(H2,84,110)(H,91,111)(H,92,117)(H,93,126)(H,94,112)(H,95,122)(H,96,125)(H,97,113)(H,98,114)(H,99,115)(H,100,119)(H,101,118)(H,102,123)(H,103,116)(H,104,120)(H,105,121)(H,106,124)(H,127,128)(H4,85,86,89)(H4,87,88,90)/t42-,43-,44-,45+,48-,49-,50-,51-,52-,53-,54-,55-,56-,57-,63-/m0/s1

InChI key

PULGYDLMFSFVBL-SMFNREODSA-N

General description

Endogenous ligand for the opioid-like G-protein coupled receptor, ORL1 (Opioid Receptor-Like 1). Has no significant activity at the µ, δ, or κ opioid receptors; however, nanomolar concentrations of Nociceptin inhibit, forskolin-stimulated adenylyl cyclase activity in cells transfected with the G-protein coupled receptor LC132, which is similar in sequence to the opioid receptor. Also reported to cause hypertension and bradycardia in mice.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Opioid-like G-protein coupled receptor, ORL1 (Opioid Receptor-Like 1)
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

H-Phe-Gly-Gly-Phe-Thr-Gly-Ala-Arg-Lys-Ser-Ala-Arg-Lys-Leu-Ala-Asn-Gln-OH

Other Notes

Madeddu, P., et al. 1999. Hypertension 33, 914.
Sandin, J., et al. 1999. NeuroReport 10, 71.
Meunier, J., et al. 1995. Nature 377, 532.
Reinscheid, R.K., et al. 1995. Science 270, 792.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danni Cao et al.
ACS chemical neuroscience, 11(19), 3036-3050 (2020-09-09)
Nalfurafine, a moderately selective kappa opioid receptor (KOR) agonist, is used in Japan for treatment of itch without causing dysphoria or psychotomimesis. Here we characterized the pharmacology of compound 42B, a 3-dehydroxy analogue of nalfurafine and compared with that of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service