Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
agency
JP, NF, Ph. Eur.
product line
EMPROVE® EXPERT
assay
10.0-10.5% (w/v), 9.5-10.0%
form
liquid
concentration
≥10 -< 20%
impurities
Residual solvents (ICH Q3C) (excluded by production process), ≤0.0001% Free chlorine (Cl), ≤0.0005% Sulfate (SO₄), ≤0.001% Sulfite (SO₃), ≤0.008% Residue on ignition (as SO₄), ≤0.01% Evaporation residue, ≤2.5 IU/mL Endotoxins
pH
<1 (20 °C in H2O)
bp
102 °C
density
1.05 g/cm3 at 20 °C
application(s)
cleaning in place
pharma/biopharma processes
storage temp.
2-25°C
SMILES string
Cl
InChI
1S/ClH/h1H
InChI key
VEXZGXHMUGYJMC-UHFFFAOYSA-N
General description
As a result, you can integrate our products directly into your biopharmaceutical production process. In addition to our comprehensive portfolio, we also offer custom manufacturing services to meet your exact needs.
Application
- To adjust the pH of process solutions, buffers, and media during upstream and downstream processing.
- In cleaning-in-place (CIP) procedures for equipment and piping, ensuring removal of residues and maintaining aseptic conditions.
Features and Benefits
- Comprehensive, up-to-date documentation you need to navigate regulatory challenges, manage risks, and improve processes.
- Suitable for higher-risk applications where the lowest microbiological and endotoxin levels are of utmost importance.
- Emprove® products are designed to facilitate compliance with global regulations (e.g., ICH Q7, EU GMP, USP, EP).
- Detailed supply chain and manufacturing information helps ensure traceability and continuity, which is critical for GMP manufacturing.
Analysis Note
- Assay (alkalimetric): 9.5 - 10.0 %:
- Assay (alkalimetric): 10.0 - 10.5 % (w/v)
- Identity: passes test
- Color: ≤ 10 Hazen
- Appearance: passes test
- Bromide or iodide: passes test
- Free bromine or chlorine: passes test
- Free chlorine (Cl): ≤ 0.0001
- Sulfate (SO₄): ≤ 0.0005 %
- Sulphite (SO3): ≤ 0.001 %
- Heavy metals (as Pb): ≤ 0.0001 %
- Residual solvents (ICH Q3C): ...excluded by production process
- Evaporation residue: ≤ 0.01
- Residue on ignition (as SO₄): ≤ 0.008 %
- Endotoxins: ≤ 2.5 I.U./ml
Filtered with 0.2 µm
Manufactured from Hydrochloric acid fuming 37% EMPROVE® ESSENTIAL PhEur, BP, JP, NF and purified water acc. PhEur, JP.
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Corresponds to Ph Eur, JP, NF
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| form liquid | form liquid | form liquid | form - |
| agency JP, Ph. Eur., NF | agency BP, ChP, JP, NF, Ph. Eur. | agency Ph. Helv. | agency - |
| density 1.05 g/cm3 at 20 °C | density 1.18 g/mL at 20 °C, 1.19 g/cm3 at 20 °C | density 1.12 g/cm3 at 20 °C, 1.18 g/mL at 20 °C | density 1.02 g/cm3 at 20 °C, 1.18 g/mL at 20 °C |
| pH <1 (20 °C in H2O) | pH <1 (20 °C in H2O) | pH <1 (20 °C in H2O) | pH <1 (20 °C) |
| bp 102 °C | bp >100 °C (lit.) | bp >100 °C (lit.), 107 °C/1013 hPa | bp >100 °C (lit.) |
| storage temp. 2-25°C | storage temp. 2-25°C | storage temp. 2-25°C | storage temp. 2-25°C |
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
12 - Non Combustible Liquids
Stay compliant with GMP using the Emprove® Program
Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.
Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.
Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite
Not finding what you are looking for? Visit the Emprove® Program Help page.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



