Sign In to View Organizational & Contract Pricing.
About This Item
Linear Formula:
C27H25ClN4O2.HCl
NACRES:
NA.28
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
solid
Quality level:
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light
Quality Level
assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, desiccated (hygroscopic), protect from light
color
beige
solubility
DMSO: 25 mg/mL
shipped in
ambient
storage temp.
−20°C
General description
A cell-permeable furanylcarboxamide compound that acts as a potent, reversible and non-ATP-competitive inhibitor of MK-2/MAPKAP-K2 activity (IC50 = 110 nM; EC50 = 350 nM for pHSP27 in IL-1β-stimulated SW1353 cells) with excellent selectivity over 150 kinases (% activity inhibition at 10 µM against human CK2, Haspin, Arg and CK1γ3 = 40, 40, 42 and >70, respectively). Shown to efficiently suppress the secretions of TNFα, IL6 (IC50 = 4.4 and 5.2 µM in LPS-stimulated THP1 cell, respectively) and MMP-13 (IC50 = 5.7 and 2.2 µM in IL1β-stimulated SW1353 and in primary osteoarthritis-derived chondrocytes, respectively). Exhibits desirable bioavailability and weakly inhibits a panel of cytochrome P450 isozymes (IC50 >20 µM for 3A4, 2D6 and 2C9).
Packaging
Packaged under inert gas
Preparation Note
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Other Notes
Huang, X., et al. 2011. ACS Med. Chem. Lett.2, 632.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Regulatory Review (Z)
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 475964-10MG | 04055977184334 |