Skip to Content
Merck
All Photos(2)

Key Documents

444158

Sigma-Aldrich

MDMX Inhibitor, NSC207895

The MDMX Inhibitor, NSC207895 controls the biological activity of MDMX. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

MDMX Inhibitor, NSC207895, 7-(4-Methylpiperazin-1-yl)-4-nitro-2,1,3-benzoxadiazole-1-oxide, 7-(4-Methylpiperazin-1-yl)-4-nitro-1-oxido-2,1,3-benzoxadiazol-1-ium, XI-006

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13N5O4
CAS Number:
Molecular Weight:
279.25
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark red

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

CN(CC1)CCN1C(C2=[N]([O-])ON=C23)=CC=C3[N+]([O-])=O

General description

A cell-permeable benzofuroxan compound that downregulates the p53 negative regulator MDMX protein level in MCF-7, LNCaP, and A549 cells (1 to 10 µM for 16 to 24 h) by suppressesing MDMX promoter transcription activity (IC50 = 2.5 µM in HT1080 cells), leading to enhanced p53 stabilization and activation. A great complement to MDM2 antagonist Nutlin-3 (Cat. Nos. 444143 & 444151) in growth inhibition and apoptosis induction of cancer cells that are more sensitive to MDMX than MDM2 inhibition.
A cell-permeable benzofuroxan compound that is shown to downregulate the p53 negative regulator MDMX protein level in MCF-7, LNCaP, and A549 cells (1 to 10 µM for 16 to 24 h) by suppressesing MDMX promoter transcription activity (IC50 = 2.5 µM in HT1080 reporter assays), leading to enhanced p53 stabilization/activation and an upregulation of the p53 downstream gene MDM2 production, which in turn could mediate further cellular MDMX proteolytic degradation. A great complement to MDM2 antagonist Nutlin-3 (Cat. Nos. 444143 & 444151) in growth inhibition of cancer cells that are more sensitive to MDMX than MDM2 inhibition, such as MCF-7 (72% vs. 42% growth inhibition by 5 µM XI-006 and Nutlin-3a, respectively, in 4 days.), where MDMX, but not MDM2, inhibition results in apoptosis induction.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wang, H., et al. 2011. Mol. Cancer Ther.7, 611.
Berkson, R.G., et al. 2005. Int. J. Cancer115, 701.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachel G Berkson et al.
International journal of cancer, 115(5), 701-710 (2005-02-25)
Activation of the p53 tumour suppressor is predicted to have therapeutically beneficial effects. Many current anti-cancer therapies activate the p53 response via DNA damage. Non-genotoxic activation of the p53 pathway would open the way to long-term and possibly prophylactic treatments.
Divya Venkatesh et al.
Genes & development, 34(7-8), 526-543 (2020-02-23)
MDM2 and MDMX, negative regulators of the tumor suppressor p53, can work separately and as a heteromeric complex to restrain p53's functions. MDM2 also has pro-oncogenic roles in cells, tissues, and animals that are independent of p53. There is less

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service