Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 1 mL | Estimated to ship TODAYfromAreál Kühne+Nagel spol. s r.o. | CZK 7,440.00 |
About This Item
CZK 7,440.00
Estimated to ship TODAYDetails
biological source
goat
Quality Level
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
liquid
contains
≤0.1% sodium azide as preservative
species reactivity
mouse
manufacturer/tradename
Calbiochem®
storage condition
do not freeze
dilution
(Enzyme Immunoassay (1:1000)
Immunoelectrophoresis )
isotype
IgG
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
General description
Application
Immunoelectrophoresis (see comments)
Packaging
Physical form
Other Notes
Dissolve 1 mg 4-methylumbelliferyl-β-D-galactoside (Cat. No. 474424) in 10 ml substrate buffer (10 mM sodium phosphate, pH 7.8, with 1 mM MgCl2) by stirring for 30 min at room temperature. Remove undissolved substrate by centrifuging or filtration.
Monospecific for mouse IgG, heavy and light chains, as determined by immunoelectrophoresis against normal mouse serum. Variables associated with assay conditions will dictate the proper working dilution.
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| species reactivity mouse | species reactivity mouse | species reactivity rabbit | species reactivity mouse |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| biological source goat | biological source goat | biological source goat | biological source goat |
| isotype IgG | isotype IgG | isotype IgG | isotype IgG |
| clone polyclonal | clone polyclonal | clone polyclonal | clone polyclonal |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 401607-1ML | 04055977211702 |



