Skip to Content
Merck
All Photos(1)

Key Documents

40-006

Millipore

BioTek® 800 TS Absorbance Reader with Gen5 software

The 800 TS easily measures absorbance in microplate-based assays. The onboard software is ideal for endpoint protocols, and under computer control with Gen5 software, applications include kinetic ELISA.

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
CZK 281,800.00

CZK 281,800.00


Estimated to ship onMay 16, 2025Details



Select a Size

Change View
1 PKG
CZK 281,800.00

About This Item

UNSPSC Code:
41110000
NACRES:
NB.51

CZK 281,800.00


Estimated to ship onMay 16, 2025Details


General description

The 800 TS Absorbance Reader is an affordable, high quality microplate reader for assays in 6- to 384-well formats. The color touchscreen provides a visual user interface, making programming simple. The onboard software includes “quick read” and custom protocols, with data viewed immediately after measure, followed by export to USB or to the compact printer. The software stores a history of programmed assays and recent plate readings for instant recall. The 800 TS can be configured to include shaking, ideal for assays like short- or long-term kinetics, enabled under computer control via Gen5 Software. The 800 TS, used alongside the 50 TS Washer, makes an affordable system to automate many application workflows, including immunoassays, cytotoxicity, enzyme assays, cell-based assays and more.

Gen5 software controls all BioTek automated microplate readers, configuring system and data collection parameters, analysis and a variety of reporting and exporting features. One-click export to Excel® makes it fast and easy to export displayed data.

Features
High quality, high performance at an affordable price
Application versatility: ELISA, protein and other endpoint assays to kinetics and cell based assays
Color touchscreen for quick, easy programming and operation
USB flash drive for convenient data export
High precision and accuracy for quality results

General Specifications
Detection modes Absorbance
Read methods Endpoint; Kinetic, well area scanning (under computer control)
Microplate types 6-, 12-, 24-, 48-, 96-well microplates
Maximum plate height 0.9" (22.86 cm)
Shaking Linear
Software Gen5 Software for external computer control and data analysis
Onboard software Up to 40 user-programmable protocols; Quick menu; Create, edit or run protocols; Save results to USB thumb drive
Printer External USB printer (optional)
User interface 4.3" color LCD touchscreen display

Absorbance Specifications
Light source Tungsten halogen
Detector Photodiode
Wavelength selection Filters
Wavelength range 400 to 750 nm
Dynamic range 0 to 4.0 OD (normal & rapid read modes); 0 to 3.0 OD (sweep read mode)
Resolution 0.0001 OD (via Gen5 control)
Filter wheel capacity 5
Filters supplied 405, 450, 490, 630
OD accuracy 96-well plate, normal read mode, 405 nm (+1.0% +0.010 OD from 0.0 to 2.0 OD)
OD linearity 96-well plate, normal read mode, 405 nm (+1.0% from 0.0 to 2.0 OD; +3.0% from 2.0 OD to 3.0 OD)
OD repeatability 96-well plate, normal read mode, 405 nm (+0.5% +0.005 OD from 0.0 to 2.0 OD)

Physical Characteristics
Connectivity (1) USB Type B for computer control and (2) USB Type A for thumb drive and printer
Power External 24VDC power supply compatible with 100-240VAC @50-60Hz
Power consumption 40 Watts
Dimensions 16.5 x 15 x 7 inches (41.9 x 38.1 x 17.8 cm)
Weight 18.5 lbs (8 kg)

Application

The 800 TS easily measures absorbance in microplate-based assays. The onboard software is ideal for endpoint protocols, and under computer control with Gen5 software, applications include kinetic ELISA.

Analysis Note

For use with ELISA. See product protocol for specific use instructions.

Other Notes

The 800 TS Absorbance Reader is an affordable, high quality microplate reader for assays in 6- to 96-well formats. The color touchscreen provides a visual user interface, making programming fast and intuitive. Applications for the 800 TS extend from endpoint ELISA to temperature sensitive kinetic assays, enabled with Gen5 Software. The 800 TS partners well with the 50 TS Washer, making a compact, versatile and affordable system to automate many application workflows.

Legal Information

BioTek is a registered trademark of BioTek/Fischer
Excel is a registered trademark of Microsoft Corporation

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service