Skip to Content
Merck

Skip To

40-092AB

BioTek® 405 TSRS Magnetic 96-well Washer

Synonym(s):

ELISA washer, MILLIPLEX® assay plate cleaner, MILLIPLEX® assay washer, MILLIPLEX® microplate washing system, MILLIPLEX® workflow plate washer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 pkg
Please contact Customer Service for Availability
CZK 500,700.00

About This Item

NACRES:
NB.51
UNSPSC Code:
41104923

CZK 500,700.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Designed to efficiently wash bead-based multiplex assays, cell-based assays, and ELISAs. Compatible with 96 well plate.

manufacturer/tradename

BioTek®

technique(s)

ELISA: suitable, multiplexing: suitable

shipped in

ambient

General description

Extended Specifications
  • Microplate Types: 96‑well plates (low profile and standard height; solid or filter bottom)
  • Residual Volume Reduction (RS): ≤ 2 μL/well for minimal carryover
  • Fluid Pump Type: Diaphragm pump
  • Instrument Control: Local touchscreen interface; PC control via Agilent BioTek LHC2 software
  • Number of Reagents: 1–4
  • Processes: Optimized cell washing, full‑plate washing, programmable shaking and soaking, Ultrasonic Advantage
  • Wash Cycles: 1–250 programmable cycles
  • Volume Range: 50–3,000 μL/well (96‑pin manifold)
  • Dispense Precision: < 3% CV at 300 μL/well (96‑well washing)
  • Wash Speed: 96‑well, 300 μL/well, 3 cycles ≤ 30 seconds
  • Flow Rates: High to low flow, optimized for cell assays
  • Washer/Dispenser Options: Vacuum filtration (optional), biomagnetic separation
The BioTek® 405 TSRS Washer is an automated microplate washing system with Residual Volume Reduction (RS) technology for minimal carryover. Ideal for ELISA and MILLIPLEX® workflows, it offers reliable performance, Ultrasonic Advantage self‑cleaning, and compatibility with multiple plate formats for high‑throughput labs.

Application

The BioTek® 405 TSRS Washer is ideal for ELISA, cell‑based assays, and MILLIPLEX® multiplex workflows. It ensures precise, automated washing with minimal residual volume for reliable, high‑throughput applications.

Features and Benefits

  • Residual Volume Reduction (RS): Ensures minimal leftover liquid in wells, improving assay accuracy and reducing background noise.
  • Verify Technology: Automated QC check for manifold tube blockages, with visual reporting of failed wells.
  • Ultrasonic Advantage: Uses ultrasonic cleaning to automatically clear aspirate and dispense manifolds, reducing maintenance.
  • Flexible Assay Support: Optimized for cell based assays, bead based multiplex assays,
  • User Interface: High resolution touchscreen with USB ports for file transfer and storage.
  • Labware Compatibility: Supports 96 well format.
  • Automation Ready: Can be integrated into automated systems for high throughput workflows.

Legal Information

BioTek is a registered trademark of BioTek Instruments LLC
Ultrasonic Advantage is a trademark of BioTek Instruments, Inc. (now part of Agilent Technologies)
Verify is a trademark of BioTek Instruments, Inc. (now part of Agilent Technologies)
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For Research Use Only. Not for use in diagnostic procedures.




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters